Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Substance P Rat, Unlabeled, Clone: NC1/34, BD

Catalog No. BDB556312 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556312 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556312 Supplier BD Biosciences Supplier No. 556312
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Peptides with transmitter-like characteristics are found in many areas of the brain, and may cause inhibition, excitation or both when applied to target neurons. These peptides are synthesized as secretory products and grouped into families according structural and functional similarity. Substance P, a member of the tachykinin family, is thought to play a role in nociceptive transmission in the dorsal horn of the spinal cord. Its proposed role is based on a number of anatomical and physiological findings. For example, substance P is present in high concentration in a subpopulation of fine-diameter primary sensory axons and in nerve terminals in dorsal horn regions where primary sensory nociceptors terminate. It specifically excites nociceptive dorsal horn and is released in the spinal cord in vivo during activation of nociceptive primary sensory fibers. Additionally, substance P receptor antagonists have been shown to block nociceptive responses in vivo and in vivo. NC1/34 is specific for substance P. It reacts with mammalian substance P and has been shown to also cross-react with a substance P-like protein in the pigeon brain. It binds to subpopulations of nerve terminals and cell bodies in the central nervous system. The initial characterization of the antibody was in rat brain. The antibody does not cross-react with other mammalian brain peptides tested including [Leu]enkephalin, [Met]enkephalin, somatostatin, or β-endorphin. NC1/34 recognizes an epitope in the carboxy terminal region of substance P and thus does not differentiate substance P from neurokinins A and B. Substance P conjugated to carrier protein was used as immunogen.

Host Species: Rat
Clone: NC1/34
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Substance P conjugated to carrier protein

Electron Microscopy, Immunohistochemistry (Frozen), Radioimmunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen Substance P
Applications Immunohistochemistry
Classification Monoclonal
Clone NC1/34
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Substance P conjugated to carrier protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Pigeon, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.