Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Substance P Rat anti-Human, Mammals, Clone: NC1, MilliporeSigma™

Catalog No. MAB356MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB356MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB356MI Supplier MilliporeSigma Supplier No. MAB356
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect Substance P using this Anti-Substance P Antibody, pain, clone NC1 validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Substance P
Applications Immunohistochemistry
Classification Monoclonal
Clone NC1
Conjugate Unconjugated
Formulation Tissue culture supernatant. Liquid containing 1mg/mL thimerosal.
Gene Accession No. NM_003182.1; NM_013996.1; NM_013997.1; NM_013998.1
Gene Symbols TAC1, NKNA, Hs.2563, NK2, PPT, NKA, TAC2
Host Species Rat
Immunogen Substance P conjugated to BSA.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.