Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Substance P Receptor Rabbit anti-Guinea Pig, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5060
Description
Anti-Substance P Receptor Antibody, pain detects level of Substance P Receptor and has been published and validated for use in immunofluoresence, immunohistochemistry and western blotting.
Specifications
Substance P Receptor | |
Polyclonal | |
Rabbit polyclonal serum in buffer containing liquid with no preservatives. | |
Rabbit | |
Unpurified | |
Primary | |
Guinea Pig, Mouse, Rat | |
Serum |
Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
TAC1; NK2; NKA; NKNA; Hs.2563; PPT; TAC2 | |
Synthetic peptide that corresponds to a 23 amino acid sequence (385-407) of the COOH terminus of the rat Substance P Receptor (NK-1). | |
50 μL | |
NM_003182.1 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction