Learn More
MilliporeSigma™ anti-Substance P Receptor, Polyclonal, ™ (Chemicon™)
Supplier: MilliporeSigma™ AB15810/DEL
Description
Substance P is involved in nociception, transmitting information about tissue damage from peripheral receptors to the central nervous system to be converted to the sensation of pain. It has been theorized that it plays a part in fibromyalgia. Capsaicin ha
Immunohistochemistry, Western Blotting
Specifications
| Substance P Receptor | |
| Unconjugated | |
| Synthetic peptide from rat Substance P Receptor. | |
| RUO | |
| Primary | |
| Serum |
| Polyclonal | |
| Guinea Pig | |
| 50 μL | |
| Neuroscience | |
| Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.