Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfotyrosine Mouse anti-All, Clone: Sulfo-1C-A2, MilliporeSigma™

Catalog No. 051100XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-110-0XMI 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-110-0XMI Supplier MilliporeSigma Supplier No. 051100X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Sulfotyrosine Antibody, Clone Sulfo-1C-A2 is a high quality Mouse Monoclonal Antibody for the detection of Sulfotyrosine and has been validated in western blot, immunoprecipitation, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sulfotyrosine
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Sulfo-1C-A2
Conjugate Unconjugated
Formulation 100μL in 100mM Tris, 150mM NaCl, 180mM Glycine, pH 7.5 containing 0.05% sodium azide.
Host Species Mouse
Immunogen This antibody was developed using a phage display library as described in Kehoe et al., 2006. The selection antigens were KAKISDP-DY(SO3)MTGYMDAC and KDKKYATEY(SO3)-EYLDYDFC
Purification Method Culture supernatant, purified by protein G affinity chromatography
Quantity 30 μL
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.