Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO-1 Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. 09409MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-409-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-409-MI Supplier MilliporeSigma Supplier No. 09409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SUMO-1 Antibody detects level of SUMO-1 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100μL of purified by salt precipitation in PBS containing 0.01% sodium azide. Dilute to working strength with 50mM Tris-HCl buffer (pH 7.6) containing 1.5% sodium chloride and 1% normal goat serum (if a goat anti-rabbit IgG linker antibody is to be us
Gene Symbols SUMO1; sentrin; Sentrin; SMT3H3; OFC10; UBL1; DAP-1; SENP2; SMT3; PIC1; GMP1; SUMO-1; SMT3C
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues 11-101 of human SUMO-1.
Purification Method Salt precipitation
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001005781.1; NM_001005782.1; NM_003352.4
Target Species All species
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.