Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SUMO1, Clone: 21C7, R&D Systems™

Catalog No. A722
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A722 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A722 Supplier R&D Systems Supplier No. A722

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Antigen SUMO1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 21C7
Conjugate Unconjugated
Dilution Western Blot 0.1-0.5 ug/mL, Immunoprecipitation
Formulation Supplied as a solution in PBS containing Glycerol and Sodium Azide. with Sodium Azide
Gene Accession No. P63165
Gene Alias DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SMT3H3, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1, UBL1
Gene Symbols SUMO1
Host Species Mouse
Immunogen Purified, recombinant human SUMO1, Accession # P63165
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7341
Test Specificity This antibody detects endogenous, human SUMOylated proteins in Western blots. Cross-reactive against murine and rat proteins.
Target Species Human
Content And Storage 6 months from date of receipt, -20 °C as supplied. 3 months, -20 °C under sterile conditions after opening.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.