Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SUMO2/3, Clone: 852908, R&D Systems™

Catalog No. A718
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A718 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A718 Supplier R&D Systems Supplier No. A718
This item is not returnable. View return policy

Rat Monoclonal Antibody

SUMO2/3 Monoclonal specifically detects SUMO2/3 in Human samples. It is validated for Western Blot.

Specifications

Antigen SUMO2/3
Applications Western Blot
Classification Monoclonal
Clone 852908
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/mL
Formulation 0.5 mg/mL in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide. with 0.09% Sodium Azide
Gene Accession No. P55854
Gene Alias HSMT3, Sentrin-2, small ubiquitin-like modifier 2, SMT3A, SMT3B, SMT3H1, SMT3H2, SUMO2, SUMO3
Gene Symbols SUMO2
Host Species Rat
Immunogen Purified, recombinant human SUMO3 Accession # P55854
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6613
Test Specificity This antibody detects endogenous, human SUMOylated proteins in Western blots. This antibody has equivalent reactivity to SUMO2 and SUMO3 in Western blots with recombinant SUMO proteins. It has less than 5% cross-reactivity with recombinant SUMO4 and no cross-reactivity with recombinant SUMO1.
Target Species Human
Content And Storage Store the unopened product at -20 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Storage below -20 degreesC is not recommended. Do not use past expiration date.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.