Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen SUMO2
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056444T
SDP
View Documents
Novus Biologicals
NB10056444SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056444
SDP
View Documents
Novus Biologicals
NB10056444
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

SUMO2 Polyclonal specifically detects SUMO2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SUMO2
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
Human, Mouse
P61956
6613
The amino acid sequences used as immunogen for this antibody are 4-16 (EKPKEGVKTENDH) of human SUMO2 (corresponding to amino acids 4-16 of SUMO3) and 61-76 (RFDGQPINETDTPAQL) of human SUMO2 (corresponding to amino acids 60-75 of SUMO3).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Epigenetics
PBS containing 0.05% BSA with 0.05% Sodium Azide
HSMT3, MGC117191, sentrin 2, Sentrin-2, small ubiquitin-like modifier 2, small ubiquitin-related modifier 2, SMT3 (suppressor of mif two 3, yeast) homolog 2, SMT3 homolog 2, SMT3 suppressor of mif two 3 homolog 2 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 2 (yeast), SMT3A, SMT3B, SMT3H2, SUMO-2, SUMO3, SUMO-3, Ubiquitin-like protein SMT3A, ubiquitin-like protein SMT3B
SUMO2
IgG
Protein G purified
Based on homology of the amino acids used as immunogen, this antibody will detect human and mouse SUMO2 and human and mouse SUMO3.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.