Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SUMO3, Human, Clone: 907614, R&D Systems™

Catalog No. A720
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A720 Supplier R&D Systems Supplier No. A720
This item is not returnable. View return policy

Rat Monoclonal Antibody

SUMO3 Monoclonal specifically detects SUMO3 in Human samples. It is validated for Western Blot.

Specifications

Antigen SUMO3
Applications Western Blot
Classification Monoclonal
Clone 907614
Conjugate Unconjugated
Dilution Western Blot
Formulation Supplied as a solution in PBS containing Glycerol and Sodium Azide. with Sodium Azide
Gene Accession No. P55854
Gene Alias SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, SMT3B, SMT3H1, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B
Gene Symbols SUMO3
Host Species Rat
Immunogen Peptide sequence from SUMO3 Accession # P55854
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6612
Test Specificity This antibody detects endogenous, human SUMOylated proteins in Western blots. This antibody detects purified, recombinant SUMO3, but has no cross-reactivity to recombinant human SUMO1, 2, or 4.
Target Species Human
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.