Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sulfite oxidase, Mouse, Clone: 4F2, Abnova™

Catalog No. 89000992 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-992 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-992 Supplier Abnova Corporation Supplier No. H00006821M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SUOX.

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins. [provided by RefSeq

Sequence: YAWSGGGRAVIRVDVSLDGGLTWQVAKLDGEEQRPRKAWAWRLWQLKAPVPAGQKELNIVCKAVDDGYNVQPDTVAPIWNLRGVLSNAWHRVHVYV

Specifications

Antigen sulfite oxidase
Applications ELISA
Classification Monoclonal
Clone 4F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SUOX.
Formulation PBS with no preservative; pH 7.4
Gene SUOX
Gene Accession No. NM_000456
Gene Symbols SUOX
Host Species Mouse
Immunogen SUOX (NP_000447, 391 a.a. ∼ 486 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6821
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.