Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superoxide Dismutase 1 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5482MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5482MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5482MI Supplier MilliporeSigma Supplier No. AB5482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Superoxide Dismutase 1 using this Anti-Superoxide Dismutase 1 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Superoxide Dismutase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50μL of sterile water.
Gene Symbols SOD1; ALS1; IPOA; Cu-Zn; SOD; ALS; homodimer; EC 1.15.1.1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 25-37 of mouse SOD1.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000454.4
Target Species Mouse, Rat
Content And Storage Lyophilized: −70°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.