Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

suppressor of Ty 16 homolog (S. cerevisiae), Mouse, Clone: 2D1, Abnova™

Catalog No. 89001207 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-207 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-207 Supplier Abnova Corporation Supplier No. H00011198M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SUPT16H.

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT (facilitates chromatin transcription), interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80kDa subunit and a 140kDa subunit; this gene encodes the 140kDa subunit. [provided by RefSeq

Sequence: PGEQTVPALNLQNAFRIIKEVQKRYKTREAEEKEKEGIVKQDSLVINLNRSNPKLKDLYIRPNIAQKRMQGSLEAHVNGFRFTSVRGDKVDILYNNIKHALFQPCDGE

Specifications

Antigen suppressor of Ty 16 homolog (S. cerevisiae)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SUPT16H.
Formulation PBS with no preservative; pH 7.4
Gene SUPT16H
Gene Accession No. NM_007192
Gene Alias CDC68/FACT/FACTP140/FLJ10857/FLJ14010/FLJ34357/SPT16/CDC68
Gene Symbols SUPT16H
Host Species Mouse
Immunogen SUPT16H (NP_009123, 608 a.a. ∼ 715 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 11198
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.