Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sur-2 Mouse anti-Human, Unlabeled, Clone: D27-1805, BD

Catalog No. BDB550429 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550429 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550429 Supplier BD Biosciences Supplier No. 550429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUR-2 (suppressor of ras) is a gene that was originally discovered in the vulval signaling pathway of the nematode Caenorhabditis elegans. Genetic studies suggest that the product of the SUR-2 gene encodes a protein Sur-2, that acts downstream of the Raf/Map kinase pathway. The human homolog of SUR-2 (hSUR-2) was shown to interact with the E1A zinc finger domain from adenovirus. The adenovirus E1A protein can bind to a number of cellular proteins to alter cell cycle progression. E1A exerts this change in the cell cycle by interacting with the transcriptional machinery of the cell. In vitro studies indicate that Sur-2 interacts with the E1A zinc finger protein and the Srb/Mediator transcriptional multiprotein complex. These studies suggest that Sur-2 may play a role as a bridge between E1A and the Srb/Mediator multiprotein complex. However, the roles of Sur-2 in regulating the transcriptional activity of the Srb/Mediator complex and E1A remain to be fully elucidated. Sur-2 has been reported to migrate in SDS/PAGE at 150kDa2, but our studies indicate that it migrates at approximately 140kDa. The antibody recognizes human Sur-2. A synthetic peptide corresponding to the N-terminus region of human Sur-2 (METQLQSIFEEVVKTEVIEC) was used as immunogen.

Host Species: Mouse
Clone: D27-1805
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human Sur-2 synthetic peptide

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sur-2
Applications Western Blot
Classification Monoclonal
Clone D27-1805
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Sur-2 synthetic peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.