Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surfactant Protein A Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3422MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3422MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3422MI Supplier MilliporeSigma Supplier No. AB3422

Goat Polyclonal Antibody

Detect Surfactant Protein A using this Anti-Surfactant Protein A Antibody validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Surfactant Protein A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS. Contains no preservative.
Gene Symbols SFTPA1; SFTP1; COLEC4; PSPA; SFTPA; PSP-A; PSAP; SP-A1; SP-A
Host Species Goat
Immunogen Human SP-A from human alveolar proteinosis lavage fluid.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001093770.1
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.