Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Surfactant Protein B Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07614
Description
Anti-Surfactant Protein B Antibody is an antibody against Surfactant Protein B for use in immunohistochemistry and western blotting.
Specifications
Surfactant Protein B | |
Polyclonal | |
rabbit antiserum containing 0.05% sodium azide before the addition of glycerol to 30% | |
Rabbit | |
Serum | |
Primary | |
−20°C for 24 months | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
SFTPB; SP-B; SFTP3; Phe; PSP-B; SFTB3; SMDP1 | |
Purified human surfactant protein B | |
50 μL | |
Human, Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction