Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surfactant Protein B Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABS21MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS21MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS21MI Supplier MilliporeSigma Supplier No. ABS21
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Surfactant Protein B Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Surfactant Protein B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Surfactant Protein B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols SFTPB; SP-B; SFTP3; Phe; PSP-B; SFTB3; SMDP1
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human Surfactant Protein B at the Saposin B-type 2 domain.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_942140
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.