Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3610MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3610MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3610MI Supplier MilliporeSigma Supplier No. AB3610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Survivin Antibody is validated for use in western blot, immunoprecipitation, immunohistochemistry for the detection of Survivin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Survivin
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with no preservatives
Gene Symbols BIRC5; IAP4; survivin; API4; SVV; EPR-1
Host Species Rabbit
Immunogen Recombinant, GST-tagged full length human Survivin
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001012270.1; NM_001012271.1; NM_001168.2
Target Species Human
Content And Storage −20°C for 6 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.