Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Survivin Mouse anti-Human, Clone: 8E2

Catalog No. MAB4617MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4617MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4617MI Supplier MilliporeSigma™ Supplier No. MAB4617

Mouse Monoclonal Antibody

Detect Survivin using this Anti-Survivin Antibody, clone 8E2 validated for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Survivin
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 8E2
Conjugate Unconjugated
Formulation Purified immunoglobulin, by Protein G. Liquid in 10mM PBS, pH 7,4 containing 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_001012270.1; NM_001012271.1; NM_001168.2
Gene Symbols BIRC5, SVV, IAP4, survivin, EPR-1, API4
Host Species Mouse
Immunogen Full length recombinant human survivin
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.