Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Survivin, Fluorescein, Clone: 91630, R&D Systems™

Catalog No. IC6472F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6472F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6472F Supplier R&D Systems Supplier No. IC6472F
This item is not returnable. View return policy

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen Survivin
Applications Flow Cytometry
Classification Monoclonal
Clone 91630
Conjugate Fluorescein
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O15392
Gene Alias API4, API4 survivin variant 3 alpha, Apoptosis inhibitor 4 baculoviral IAP repeat-containing protein 5, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, BIRC5, EPR-1, IAP4
Gene Symbols BIRC5
Host Species Mouse
Immunogen E. coli-derived recombinant human Survivin, Met1-Asp142, Accession # O15392
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 332
Test Specificity Detects human Survivin in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.