Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUV39H1 Mouse anti-Human, Mouse, Clone: MG44, MilliporeSigma™

Catalog No. 05615MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-615-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-615-MI Supplier MilliporeSigma Supplier No. 05615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SUV39H1 Antibody, clone MG44 is a Mouse Monoclonal Antibody for detection of SUV39H1 also known as KMT1A, Histone H3-K9 Methyltransferase 1 and has been validated in Chimmunoprecipitation, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUV39H1
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MG44
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative.
Gene Accession No. NM_003173.2
Gene Symbols SUV39H1, Su, SUV39H, MG44
Host Species Mouse
Immunogen MBP fusion protein corresponding to residues 1-412 of human SUV39H1
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.