Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Suv4–20h2 Rat anti-Human, Clone: 6B9, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABE369
Description
Detect the Suv4–20h2 protein using this Anti-Suv4–20h2, clone 6B9 validated for use in western blotting.
Specifications
Suv4-20h2 | |
Monoclonal | |
Unconjugated | |
NP_116090 | |
Rat | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Western Blot | |
6B9 | |
Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
SUV420H2, KMT5C, PP7130 | |
GST-tagged recombinant protein corresponding to human Suv4-20h2. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction