Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 Large T Antigen Mouse, Unlabeled, Clone: PAB 100, BD

Catalog No. BDB554154 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554154 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554154 Supplier BD Biosciences Supplier No. 554154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Simian virus 40 is a small DNA virus encoded by 5.2 kb of double-stranded DNA. SV 40 large T antigen (T-ag) is a multifunctional 85kda phosphoprotein, which is the sole viral protein required for SV40 replication. All other factors are provided by the infected host cell. In addition to its role in SV40 DNA replication, T-ag also causes transformation of susceptible cell lines. Studies of various mutant T-ag proteins have shown that the replication and transformation fractions of T-ag can be separated. The multifunctional nature of this protein has resulted in its use as a model system in a wide variety of disciplines. SV40 T-ag exercises negative regulation on the transcription of SV40 early mRNA by feedback inhibition and exerts positive regulation on transcription from the late promoter. In addition to transcriptional regulation, T-ag is involved in viral DNA replication. Specific biochemical functions required for DNA synthesis that are inherent to the T-ag include high-affinity binding to sites within the viral origin of DNA synthesis and ATPase and helicase activities. Other functions attributed to T-ag include cellular transformation, induction of cellular DNA synthesis, induction of rRNA synthesis, and provision of a host-range function for viral replication. However, all functions of T-ag are influenced by a wide range of post-translational modifications including phosphorylation, glycosylation, acetylation, acylation, and adenylation. T-ag exists in monomeric as well as polymeric forms and associates with the tumor suppressor proteins p53 and Rb (retinoblastoma protein). Most of T-ag is transported to the nucleus, while a small fraction is localized at the cell surface. PAb 100 recognizes an epitope between amino acids 270 and 520 of T-ag. PAb 100 was originally known as clone 412. Studies have suggested that PAb 100 binds the strongest to newly synthesized T-ag and to ATPase-active T-ag in some experimental systems. In other experimental systems it appears to preferentially bind to recognize mature T-ag. PAb 100 (i.e., clone 412) was developed along with a panel of monoclonal antibodies where SV40-transformed BALB/c mouse cell lines (SVT2 or B4) were used as immunogens. The specificity of the antibody was originally characterized by a variety of techniques using SV40-infected and SV40-transformed cells.

Host Species: Mouse
Clone: PAb 100
Isotype: IgG1
Species Reactivity: Viral-transformed cells
Immunogen: SV40-transformed BALB/c mouse cell lines
Formula Weight [Chemical]: 85-100kDa

Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen SV40 Large T Antigen
Applications Immunoprecipitation
Classification Monoclonal
Clone PAB 100
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen SV40-transformed BALB/c mouse cell lines
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Virus
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.