Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-SV40 T Antigen Clone: PAb280,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ DP14L100UG
Description
Specifically detects SV40 T Antigen Clone: PAb280 in SV40 infected cells samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting
Both the SV40 large T and small T antigens are encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular transforming protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the p105 product of the retinoblastoma susceptibility gene.
Specifications
SV40 T Antigen | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Simian virus 40 (SV40) | |
Purified |
Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
PAb280 | |
Lyophilized from a volatile buffer, 100μg BSA. | |
the SV40 small T-antigen | |
RUO | |
Recognizes the ∽21kDa SV40 small T-antigen in SV80 cells. Does not cross-react with SV40 large T-antigen. | |
2°C to 8°C | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction