Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SWAP-70 protein, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SWAP70 protein.
Supplier: Abnova Corporation H00023075D01P
Description
Sequence: MGSLKEELLKAIWHAFTALDQDHSGKVSKSQLKVLSHNLCTVLKVPHDPVALEEHFRDDDEGPVSNQGYMPYLNRFILEKVQDNFDKIEFNRMCWTLCVKKNLTKNPLLITEEDAFKIWVIFNFLSEDKYPLIIVSEEIEYLLKKLTEAMGGGWQQEQFEHYKINFDDSKNGLSAWELIELIGNGQFSKGMDRQTVSMAINEVFNELILDVLKQGYMMKKGHRRKNWTERWFVLKPNIISYYVSEDLKDKKGDILLDENCCVESLPDKDGKKCLFLVKCFDKTFEISASDKKKKQEWIQAIHSTIHLLKLGSPPPHKEARQRRKELRKKQLAEQEELERQMKELQAANESKQQELEAVRKKLEEAASRAAEEEKKRLQTQVELQARFSTELEREKLIRQQMEEQVAQKSSELEQYLQRVRELEDMYLKLQEALEDERQARQDEETVRKLQARLLEEESSKRAELEKWHLEQQQAIQTTEAEKQELENQRVLKEQALQEAMEQLEQLELERKQALEQYEEVKKKLEMATNKTKSWKDKVAHHEGLIRLIEPGSKNPHLITNWGPAAFTEAELEEREKNWKEKKTTESpecifications
SWAP-70 protein | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human SWAP70 protein. | |
SWAP70 | |
FLJ39540/HSPC321/KIAA0640/SWAP-70 | |
Rabbit | |
Affinity Purified | |
RUO | |
23075 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_015055.2 | |
SWAP70 | |
SWAP70 (NP_055870.2, 1 a.a. ∼ 585 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction