Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syncollin, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018705 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-705 Supplier Abnova Corporation Supplier No. H00342898D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SYCN protein.

Sequence: MSPLRPLLLALALASVPCAQGACPASADLKHSDGTRTCAKLYDKSDPYYENCCGGAELSLESGADLPYLPSNWANTASSLVVAPRCELTVWSRQGKAGKTHKFSAGTYPRLEEYRRGILGDWSNAISALYCRCS

Specifications

Antigen syncollin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SYCN protein.
Formulation PBS with no preservative; pH 7.4
Gene SYCN
Gene Accession No. XM_371167.4
Gene Alias FLJ27441/INSSA1/SYL
Gene Symbols SYCN
Host Species Rabbit
Immunogen SYCN (NP_001073937.1, 1 a.a. ∼ 134 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342898
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.