Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syk Mouse anti-Human, FITC, Clone: 4D10, BD

Catalog No. BDB552476 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552476 Supplier BD Biosciences Supplier No. 552476
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Syk is a member of the family of non-receptor type protein-tyrosine kinases and plays a crucial role in lymphocyte signaling and development. Syk is expressed in all haematopoietic cells and contributes to the signal transduction process by binding to a tyrosine-based activation motif (ITAM) of immune receptors, including Igα, TCRζ, CD3ε, FcεRIβ, and FceRIγ. Upon receptor activation, Syk binds to phosphorylated ITAMs via its two N-terminal SH2 domains thereby activating Syk and causing tyrosines in Syk to undergo auto-phosphorylation or phosphorylation. These phosphorylated sites can act as binding sites for other signaling molecules or help to regulate enzymatic activity. For example, in mast cells, Syk can activate downstream targets such as phospholipase Cγ1 and VAV. Studies in Syk-/- mast cells identified defects in both the ERK-MAP and JNK-MAP kinase pathways. Syk migrates as 72kDa in SDS/PAGE. (Predicted MW from SWISS-PROT is 72kDa; Accession number P43405).

Host Species: Mouse
Clone: 4D10
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Human Syk peptide aa. 314-339

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Syk
Applications Flow Cytometry
Classification Monoclonal
Clone 4D10
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Syk peptide aa. 314-339
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.