Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYK Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001796
Description
SYK Polyclonal specifically detects SYK in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
SYK | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_003168.2; NP_001128524.1). Reported pairs of variants represent identical proteins (NP_003168.2 and NP_001167638.1; NP_001128524.1 and NP_001167639.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_003168.2 | |
SYK | |
Peptide with sequence C-KYLQQNRHVKDKN corresponding to internal region according to NP_001128524.1. | |
0.1 mg | |
Immunology, Innate Immunity, Mast Cell Markers, Protein Kinase | |
6850 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction