Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Synapsin-1 Clone: MEGS3-10A,

Catalog No. MABN1847MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1847MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1847MI Supplier MilliporeSigma™ Supplier No. MABN1847
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Synapsin-1 Clone: MEGS3-10A in Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Synapsin-1 (UniProt O88935; also known as Synapsin I) is encoded by the Syn1 (also known as Syn-1) gene (Gene ID 20964) in murine species. The synapins constitute a family of abundant neuronal phosphoproteins that regulate SV trafficking and neurotransmitter release at the pre-synaptic terminal. Three genes exisit in mammals, encoding altogether 11 synapsin members by alternative splicings (Synapsin Ia & Ib by SYN1, Synapsin IIa & IIb by SYN2, Synapsin IIIa through IIIf by SYN3), Syn III is the most precociously expressed isoform that has a role in the early phases of neural development and is downregulated in mature neurons. On the other hand, Syn I and Syn II are expressed at low levels at birth and their expressions progressively increase during synaptogenesis to reach a stable plateau at 1-2 months of life. The NH2-terminal region is divided in A, B, and C domains and is highly conserved among synapsin isoforms. Synapsins are regulated by postransloational phosphorylations. Domain A contains PKA and CaMKI/IV phosphorylation sites, domain B contains MAPK/Erk phosphorylation sites, and domain C is phosphorylated by Src. The c-terminal regions contain spliced domains and diverge among synapsin isoforms (domain D in Syn Ia and Ib, domain G in Syn Iia and Iib, domain H in Syn Iia, and domain J in Syn IIIa), although they all bear proline-rich regions binding to several SH3-containing proteins and additional phosphorylation sites for CaMKII, MAPK/Erk, and cdk1/5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synapsin-1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone MEGS3-10A
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O88935
Gene Symbols Syn1, Syn-1
Host Species Mouse
Immunogen Recombinant full-length mouse synapsin-1.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_038708.3
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.