Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin I Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1543PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1543PMI 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1543PMI Supplier MilliporeSigma Supplier No. AB1543P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Synapsin I Antibody is validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation, western blotting for the detection of Synapsin I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synapsin I
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer lyophilized from 5mM ammonium bicarbonate. Reconstitute with 50μL of PBS.
Gene Symbols SYN1; SYN1a; SYNI; SYN1b
Host Species Rabbit
Immunogen Synapsin I (mixture of Ia & Ib) purified from bovine brain.
Purification Method Immunoaffinity purified
Quantity 10 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006950.3
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.