Learn More
Synapsin I Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Synapsin I |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal antiserum in buffer containing no preservatives. |
| Gene Symbols | SYN1; SYN1a; SYNI; SYN1b |
| Host Species | Rabbit |
| Immunogen | Synapsin I (mixture of Ia & Ib) purified from bovine brain. |
| Purification Method | Unpurified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.