Learn More
Description
Specifications
Specifications
| Antigen | Synapsin I, phosphoSer 603 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin presented as a liquid in 10mM HEPES, pH 7.5, with 150mM NaCl, 100μg/mL BSA and 50% glycerol. |
| Gene Symbols | SYN1; SYN1a; Synapsin-1; SYNI; SYN1b |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids surrounding the phosphoSer603 of rat Synapsin I. The immunogen sequence is identical in human, mouse and Xenopus. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.