Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptobrevin Mouse anti-Human, Hamster, Pig, Rat, Salamander, Clone: SP10, MilliporeSigma™

Catalog No. MAB333MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB333MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB333MI Supplier MilliporeSigma Supplier No. MAB333
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Synaptobrevin Antibody, clone SP10 is an antibody against Synaptobrevin for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptobrevin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SP10
Conjugate Unconjugated
Formulation Liquid, contains sodium azide.
Gene Accession No. NM_014231.3; NM_016830.2; NM_199245.1
Gene Symbols VAMP1, SYB1, Synaptobrevin-1, VAMP-1, DKFZp686H12131
Host Species Mouse
Immunogen Crude synaptic immunoprecipitate (Human).
Quantity 100 μL
Primary or Secondary Primary
Target Species Hamster, Human, Pig, Rat, Salamander
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.