Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptobrevin Mouse anti-Human, Pig, Feline, Clone: SP11, MilliporeSigma™

Catalog No. MAB335MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB335MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB335MI Supplier MilliporeSigma Supplier No. MAB335
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Synaptobrevin Antibody, clone SP11 is validated for use in ELISA, western blot, immunohistochemistry (paraffin) for the detection of Synaptobrevin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptobrevin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SP11
Conjugate Unconjugated
Formulation Liquid in 0.02 MPB and 0.25M NaCl, PH 7.6 with 0.1% sodium azide.
Gene Accession No. NM_014231.3; NM_016830.2; NM_199245.1
Gene Symbols VAMP1, SYB1, Synaptobrevin-1, VAMP-1, DKFZp686H12131
Host Species Mouse
Immunogen Crude synaptic immunoprecipitate (Human)
Quantity 100 μL
Primary or Secondary Primary
Target Species Feline, Human, Pig
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.