Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptogyrin 2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001490
Description
Synaptogyrin 2 Polyclonal specifically detects Synaptogyrin 2 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
Synaptogyrin 2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
cellugyrin, MGC102914, synaptogyrin 2, synaptogyrin-2 | |
Goat | |
Affinity Purified | |
RUO | |
9144 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_004701. | |
SYNGR2 | |
Peptide with sequence CIYGEGYSNAHESKQ corresponding to internal region according to NP_004701.1. | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Mouse, Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction