Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptojanin 1 Mouse, Unlabeled, Clone: 26, BD

Catalog No. BDB612249 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612249 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612249 Supplier BD Biosciences Supplier No. 612249

Mouse Monoclonal Antibody

Phosphoinositide turnover is a mechanism of intracellular signaling that involves phosphorylation of the inositol ring to produce signaling molecules that include phosphoinositol (PI)-3-P, PI-3,4-P2, PI-3,4,5-P3, PI-4-P, and PI-4,5-P2. These moleclules function in many signaling pathways to regulate events at cell membranes, such as endocytosis, exocytosis, and cell morphology. Synaptojanin 1 is a phosphoinositide phosphatase that selectively cleaves the 3-, 4-, and 5- phosphates from PIs. Two splice variants of synaptojanin 1 include a 145kDa form that is found in adult brain, and a 170kDa that is widely expressed in non- neuronal cells. The 145kDa form contains a Sac1 domain that has phosphoinositide phosphatase activity, a second phosphatase domain that has only PI-5 phosphatase activity (PI-5), and a proline-rich SH3-binding domain. The 170kDa form contains these domains, as well as a second SH3-binding consensus sequence. Synaptojanin binds many proteins implicated in endocytosis, including amphiphysins and endophilin. Thus, synaptojanin 1 may regulate endocytosis through modification of the interactions between SH3-domain proteins and PIs.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptojanin 1
Applications Western Blot
Classification Monoclonal
Clone 26
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat Synaptojanin 1 aa. 1145-1259
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.