Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Synaptophysin Clone: SP15,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 57382250UL
Description
Specifically detects Synaptophysin Clone: SP15 in Bovine, Feline, Hamster, Human, Mouse, Porcine, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting
Synaptophysin is a ∽38kDa integral membrane protein on synaptic vesicles. This protein consists of four transmembrane domains, with its amino- and carboxyl-terminus facing the cytoplasm. Due to its transmembrane domain structure, synaptophysin has been postulated to be a gap junction-like protein as voltage-dependent channel activity is seen when synaptophysin hexamers are reconstituted into lipid membranes. Physiological evidence for synaptophysin’s role in secretion includes its involvement in calcium-dependent glutamate secretion in Xenopus oocytes injected with rat cerebellum mRNA.
Specifications
Synaptophysin | |
Monoclonal | |
Unconjugated | |
Mouse | |
50 μL | |
Primary | |
Bovine, Feline, Hamster, Human, Mouse, Pig, Rat | |
Purified |
Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
SP15 | |
In PBS, 50% glycerol, pH 7.2. | |
synaptic vesicle-containing fractions from human brain homogenates | |
RUO | |
Recognizes the ∽38kDa synaptophysin protein in brain tissue. Does not cross-react with the ∽65kDa form in mouse liver extracts. | |
−20°C | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction