Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9272MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9272MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9272MI Supplier MilliporeSigma Supplier No. AB9272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Synaptophysin Antibody detects level of Synaptophysin and has been published and validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptophysin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 10mM PBS, pH 7.4 containing BSA and 0.1% sodium azide.
Gene Symbols SYP; synaptophysin; Synaptophysin
Host Species Rabbit
Immunogen Synthetic peptide from human synaptophysin.
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003179.2
Target Species Human, Rat
Content And Storage 2-8°C for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.