Learn More
Synaptotagmin Mouse, Unlabeled, Clone: 41, BD
Mouse Monoclonal Antibody
Specifications
Antigen | Synaptotagmin |
---|---|
Clone | 41 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB610433
|
BD Biosciences
610433 |
50 μg | N/A | N/A | N/A | ||||
Description
Synaptotagmin (p65) is an abundant synaptic vesicle protein that contains a single transmembrane region and two copies of an internal repeat that is homologous to the regulatory region of Protein Kinase C. It appears that synaptotagmin has a regulatory role in the synaptic vesicle pathway, particularly in vesicle docking and/or fusion with the plasmalemma. A model has been proposed to explain docking, activation, and fusion of synaptic vesicles with donor membranes. This model suggests that VAMP/synaptobrevin and synaptotagmin (vSNARE) on the synaptic vesicle, and SNAP-25 and syntaxin (tSNAREs) on the plasma membrane, interact to form a 7S complex. Two additional soluble proteins, αSNAP and NSF, are later added to the 7S complex, accompanied by the loss of synaptotagmin. The resulting 20S complex contains syntaxin, SNAP-25, VAMP, αSNAP, and NSF. Genetic studies in several species demonstrate that mutation or deletion of synaptotagmin results in a large decrease in Ca2+ triggered transmitter release. Mammalian synapses that lack synaptotagmin show a selective decrease in a fast component of release, suggesting that synaptotagmin is the Ca2+ sensor triggering exocytosis.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Synaptotagmin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
41 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.