Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin I Mouse anti-Bovine, Chicken, Mouse, Rat, Rabbit, Frog, Shark, MilliporeSigma™

Catalog No. MAB5200MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5200MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5200MI Supplier MilliporeSigma Supplier No. MAB5200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Synaptotagmin I Antibody is validated for use in immunoprecipitation, western blot, IC for the detection of Synaptotagmin I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptotagmin I
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS with 50% glycerol and 0.09% sodium azide.
Gene Accession No. NM_005639.1
Gene Symbols SYT1, DKFZp781D2042, SVP65, SytI, SYT, p65, P65, Synaptotagmin-1
Host Species Mouse
Immunogen Rat brain synaptotagmin.
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Frog, Mouse, Rabbit, Rat, Shark
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.