Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-1 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB8230
Description
Anti-Syndecan-1 Antibody is an antibody against Syndecan-1 for use in western blotting.
Specifications
Syndecan-1 | |
Polyclonal | |
Rabbit serum containing 0.05% NaN3. | |
Rabbit | |
100 μL | |
NM_001006946.1 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Western Blot | |
Unconjugated | |
CD138 ; SDC; SYND1 ; syndecan ; OTTHUMP00000115942 | |
His-tagged, full-length human recombinant Syndecan-1 protein. | |
Primary | |
Human, Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction