Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-4 Rat anti-Mouse, PE, Clone: KY/8.2, BD

Catalog No. BDB550352 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550352 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550352 Supplier BD Biosciences Supplier No. 550352
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The KY/8.2 mAb reacts with mouse syndecan-4, a cell-surface heparan sulfate proteoglycan. Although the ligand(s) for this molecule is unknown, syndecans are known to interact with the heparin-binding domain of growth factors, chemokines, and extracellular matrix components. Accordingly, heparin inhibits the binding of syndecan-4 to the pre-B cell line BC7.12. Syndecan-4 is localized to focal adhesions and binds protein-kinase Cα. Thus, it is involved in cytoskeletal organization and signaling. Syndecan-4 is expressed on stromal cells and B-lymphocyte precursors in bone marrow, in addition to most splenic B cells and a small population of T lymphocytes. It is highly expressed on activated B lymphocytes, but is downregulated in isotype-switched memory B cells. The ectodomain of syndecan-4 is shed from cell surfaces constitutively and in response to specific physiological signals, and the resulting soluble fragments are functional. Immobilized KY/8.2 mAb induced the formation of dendritic processes on syndecan-4-transfected cells and on anti-CD40-activated splenic B cells. Other methods of activating B cells have not produced the same effect. This antibody recognizes an epitope in the N-terminus of syndecan-4, regardless of post-translational modifications.

Host Species: Rat
Clone: KY/8.2
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BCB10 mouse pre-B cell line

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan 4
Applications Flow Cytometry
Classification Monoclonal
Clone KY/8.2
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BCB10 mouse pre-B cell line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.