Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SynGAP Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. 06900MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-900-MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-900-MI Supplier MilliporeSigma Supplier No. 06900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SynGAP Antibody detects level of SynGAP and has been published and validated for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SynGAP
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% NaN3 and 30% Glycerol
Gene Symbols SYNGAP1; RASA5; DKFZp761G1421; OTTHUMP00000064825; SYNGAP; RASA1; KIAA1938
Host Species Rabbit
Immunogen peptide (KRLLDAQRGSFPPWVQQTRV) corresponding to amino acids 1274-1293 of rat SynGAP-a
Purification Method Protein A purfied
Quantity 1 Unit
Primary or Secondary Primary
Gene ID (Entrez) NM_006772.1
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.