Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin-3 Mouse anti-Mouse, Rat, Human, Clone: 1-146, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2258
Description
Detect Syntaxin-3 using this Anti-Syntaxin-3 Antibody, clone 1-146 validated for use in western blot, immunoprecipitation, immunohistochemistry.
Specifications
Syntaxin-3 | |
Monoclonal | |
Unconjugated | |
NP_112386 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for up to 12 months. | |
IgG1 κ |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1-146 | |
Purified mouse monoclonal in buffer containing 0.1MTris-glycine, pH 7.4, 150mM NaCl with 0.05% NaN3. | |
STX3A | |
GST-tagged recombinant protein corresponding to residues contained within rat Syntaxin-3. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction