Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin-3 FITC Mouse anti-Human, FITC, Clone: 1-146, MilliporeSigma™

Catalog No. FCMAB155FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB155FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB155FMI Supplier MilliporeSigma Supplier No. FCMAB155F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Detect Syntaxin-3 using this Milli-Mark™ Anti-Syntaxin-3 FITC Antibody, clone 1-146 validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin-3
Applications Flow Cytometry
Classification Monoclonal
Clone 1-146
Conjugate FITC
Formulation Purified mouse monoclonal IgG1κ conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_112386
Gene Symbols STX3A
Host Species Mouse
Immunogen This antibody is generated from rabbits immunized with a GST-tagged recombinant protein corresponding to residues contained within rat Syntaxin-3.
Purification Method Protein G purfied
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.