Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin 4 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5330200UL
Description
This Anti-Syntaxin 4 Antibody is validated for use in western blotting for the detection of Syntaxin 4.
Specifications
Syntaxin 4 | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.025% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
Western Blot | |
Unconjugated | |
STX4; p35-2; STX4A; Syntaxin-4 | |
Highly purified corresponding to residues 2-23 of rat or mouse Syntaxin 4 (Accession Q08850). | |
200 μL | |
NM_004604.3 | |
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction