Learn More
Description
Specifications
Specifications
| Antigen | Syntaxin 4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.025% sodium azide. Reconstitute with 50μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). |
| Gene Symbols | STX4; p35-2; STX4A; Syntaxin-4 |
| Host Species | Rabbit |
| Immunogen | Highly purified corresponding to residues 2-23 of rat or mouse Syntaxin 4 (Accession Q08850). |
| Purification Method | Affinity Purified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.