Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 6 Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB610635 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610635 50 μg
BDB610636 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610635 Supplier BD Biosciences Supplier No. 610635
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Signal transmission between cells is often regulated by the secretion of neurotransmitters or hormones into the extracellular space. This process is controlled by a complex pathway of membrane trafficking which leads to membrane fusion and secretion. Syntaxin 1 is involved in three important complexes that modulate the secretion process: syntaxin and n-sec-1, syntaxin, VAMP, and SNAP-25, and syntaxin, VAMP, SNAP-25, αSNAP, and NSF (20S complex). Syntaxin 6 is a 255 amino acid member of the syntaxin family. It contains a C-terminal transmembrane domain and is located at the Golgi apparatus. In addition, Syntaxin 6 displays significant (47% identity) homology with SNAP-25. In vitro, Syntaxin 6 has been reported to interact with α-SNAP. Syntaxin 6 has been reported to have a calculated molecular weight of 30.6kDa, but has been observed to migrate in a range inbetween ∽ 31-35kDa.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 6
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.