Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 8 Mouse, Unlabeled, Clone: 48, BD

Catalog No. BDB611352 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611352 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611352 Supplier BD Biosciences Supplier No. 611352
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Eukaryotic protein trafficking involves the packaging of molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Recognition between vesicle and acceptor membrane is mediated by the pairing of the integral membrane SNARE proteins. The stable interaction between vesicle proteins (v-SNAREs) and target proteins (t-SNAREs) juxtaposes the membranes and results in an activated docking state and/or membrane fusion. The syntaxin protein family contains a number of members that serve as functional t-SNAREs. One member of this family, syntaxin 8, has two coiled-coil domains, one in the N-terminus and one toward the C-terminus. The C-terminal domain is homologous to a similar domain in syntaxin 6 and is predicted to interact in the formation of SNARE complexes, while a hydrophobic domain in the C-terminus may be involved in membrane anchoring. Syntaxin-8 is ubiquitously expressed, but is present at high levels in the heart and localizes specifically to the ER. Thus, syntaxin-8 is a t-SNARE that is thought to be involved in the early secretory pathway of many different cell types.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 8
Applications Western Blot
Classification Monoclonal
Clone 48
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Syntaxin 8 aa. 57-164
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.