Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synuclein α Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5038MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5038MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5038MI Supplier MilliporeSigma Supplier No. AB5038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Synuclein Antibody, α detects level of Synuclein and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synuclein α
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum lyophilized, no preservatives. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any residue.
Gene Symbols SNCA; PD1; NACP; PARK4; MGC110988; PARK1
Host Species Rabbit
Immunogen Human alpha-synuclein residue 111-131 coupled to KLH.
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000345.2
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.